Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010049.3 | NP_034179.1 | 6 | 1000 | ||
GenBank mRNA | AK031241.1 | - | 6 | 634 | ||
AK136212.1 | - | 6 | 1058 | |||
BC005796.1 | - | 6 | 572 | 3707582 | ||
L26316.1 | - | 6 | 579 | |||
V00731.1 | - | 625 | ||||
V00734.1 | - | 6 | 636 | |||
X56066.1 | - | 6 | 546 |