Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010054.2 | NP_034184.1 | 3 | 1359 | ||
GenBank mRNA | AK144652.1 | - | 3 | 1360 | ||
BC094317.1 | - | 3 | 1059 | 5718422 | ||
M80540.1 | - | 3 | 1297 | |||
M83184.1 | - | 3 | 922 |