Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF002985.1 | - | 4 | 411 | ||
AF030514.1 | - | 4 | 393 | |||
AL703031.1 | - | 4 | 370 | |||
BC005292.1 | - | 4 | 384 | 3934139 | ||
BC012532.1 | - | 4 | 538 | 4096572 | ||
BC110986.1 | - | 4 | 393 | 5178174 | ||
U59286.1 | - | 4 | 349 |