Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010086.4 | NP_034216.3 | 2 | 1022 | ||
GenBank mRNA | AF167402.1 | - | 1 | 1010 | ||
AK029528.1 | - | 2 | 1023 | |||
AK133199.1 | - | 2 | 1038 | |||
BC145785.1 | - | 1 | 911 | 40131088 |