Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113414.1 | NP_001106885.1 | 4 | 909 | ||
NM_001113415.1 | NP_001106886.1 | 4 | 909 | |||
NM_010096.3 | NP_034226.1 | 4 | 909 | |||
GenBank mRNA | AK220542.1 | - | 4 | 909 | ||
BC067018.1 | - | 5 | 664 | 6833120 | ||
U82442.1 | - | 3 | 206 | |||
U92702.1 | - | 4 | 450 | |||
U92704.1 | - | 4 | 450 |