Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010100.3 | NP_034230.1 | 12 | 2005 | ||
GenBank mRNA | AF160502.1 | - | 12 | 2005 | ||
AK004576.2 | - | 6 | 1134 | |||
BC068315.1 | - | 12 | 1963 | 30098537 |