Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013470.2 | NP_038498.2 | 6, 7 | 615 | ||
GenBank mRNA | AJ001633.1 | - | 6, 7 | 598 | ||
AK013850.1 | - | 4, 5 | 368 | |||
AK090055.1 | - | 6, 7 | 633 | |||
AK150922.1 | - | 6, 7 | 616 | |||
AK150951.1 | - | 6, 7 | 583 | |||
AK151771.1 | - | 6, 7 | 575 | |||
AK152837.1 | - | 6, 7 | 542 | |||
AK152863.1 | - | 6, 7 | 603 | |||
AK166389.1 | - | 6, 7 | 587 | |||
AK168396.1 | - | 6, 7 | 511 | |||
AK168507.1 | - | 6, 7 | 543 | |||
AK169113.1 | - | 6, 7 | 491 | |||
AK169423.1 | - | 6, 7 | 497 | |||
BC090634.1 | - | 6, 7 | 609 | 30942562 | ||
BC141167.1 | - | 6, 7 | 569 | 9055779 | ||
CT010280.1 | - | 4, 5 | 398 |