Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013482.2 | NP_038510.2 | 4 | 420 | ||
GenBank mRNA | BC053392.1 | - | 4 | 356 | 30075322 | |
L08967.1 | - | 4 | 396 | |||
L10627.1 | - | 4 | 425 | |||
L29788.1 | - | 4 | 392 |