Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289470.1 | NP_001276399.1 | 9 | 1233 | ||
NM_013489.3 | NP_038517.1 | 9 | 1236 | |||
GenBank mRNA | AF043445.1 | - | 8 | 1111 | ||
AK154834.1 | - | 8 | 1081 |