Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013491.2 | NP_038519.1 | 6, 7 | 867 | ||
GenBank mRNA | AK036735.1 | - | 6, 7 | 869 | ||
AY046403.1 | - | 6, 7 | 692 | |||
AY046404.1 | - | 7, 8 | 771 | |||
BC114336.1 | - | 6, 7 | 832 | 40048966 |