Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bhmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016668.3 | NP_057877.1 | 8 | 1886 | ||
GenBank mRNA | AK077108.1 | - | 8 | 1694 | ||
AK145668.1 | - | 8 | 1728 | |||
AK166754.1 | - | 8 | 1695 | |||
AK166818.1 | - | 8 | 1694 | |||
BC037004.1 | - | 8 | 1688 | 5100429 | ||
BC037264.1 | - | 1 | 480 | |||
BC093510.1 | - | 8 | 1661 | 5101184 | ||
BC110307.1 | - | 8 | 2089 | 5310035 |