Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Auh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016709.2 | NP_057918.2 | 9 | 879 | ||
GenBank mRNA | AK003219.1 | - | 9 | 859 | ||
BC026525.1 | - | 9 | 817 | |||
BC049597.1 | - | 9 | 863 | 6593786 |