Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081249.1 | NP_001074718.1 | 8 | 4615 | ||
NM_019389.2 | NP_062262.2 | 7 | 1735 | |||
GenBank mRNA | AK014525.1 | - | 7 | 1545 | ||
D16263.1 | - | 6 | 1295 | |||
D28599.1 | - | 8 | 4347 |