Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020332.4 | NP_065065.3 | 4, 5 | 851 | ||
GenBank mRNA | AF001533.2 | - | 2, 3 | 197 | ||
AF274752.1 | - | 4, 5 | 554 | |||
AK083135.1 | - | 4, 5 | 852 | |||
AK148070.1 | - | 4, 5 | 624 | |||
BC042644.1 | - | 4, 5 | 776 | |||
BC054379.1 | - | 4, 5 | 758 | 6390764 | ||
DQ832285.1 | - | 4, 5 | 851 |