Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301230.1 | NP_001288159.1 | 3 | 449 | ||
NM_001301232.1 | NP_001288161.1 | 2 | 342 | |||
NM_023635.6 | NP_076124.1 | 2 | 300 | |||
GenBank mRNA | AB046693.1 | - | 1 | 98 | ||
AB120511.1 | - | 1 | 76 | |||
AB232622.1 | - | 1 | 76 | |||
AF304376.1 | - | 2 | 255 | |||
AK010373.1 | - | 2 | 218 | |||
AK030529.1 | - | 2 | 247 | |||
BB857060.1 | - | 2 | 318 | |||
BC008173.1 | - | 3 | 406 | 3710213 | ||
CJ077071.1 | - | 2 | 300 | |||
CV784419.1 | - | 2 | 211 |