Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GZMM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258351.1 | NP_001245280.1 | 5 | 914 | ||
NM_005317.3 | NP_005308.1 | 5 | 918 | |||
GenBank mRNA | BC025701.1 | - | 5 | 890 | 5222281 | |
L23134.1 | - | 5 | 880 |