Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301164.1 | NP_001288093.1 | 5 | 965 | ||
NM_001301165.1 | NP_001288094.1 | 5 | 533 | |||
NM_053070.3 | NP_444300.1 | 5 | 560 | |||
GenBank mRNA | AF291660.1 | - | 4 | 455 | ||
AK090390.1 | - | 5 | 564 | |||
AK144275.1 | - | 5 | 534 | |||
AY075021.1 | - | 5 | 561 | |||
BC038270.1 | - | 5 | 940 | |||
BC094913.1 | - | 5 | 548 | 30016726 |