Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRDT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242805.2 | NP_001229734.2 | 11 | 1901 | ||
NM_001242806.2 | NP_001229735.2 | 10 | 1844 | |||
NM_001242807.2 | NP_001229736.2 | 9 | 1500 | |||
NM_001242808.2 | NP_001229737.2 | 9 | 1694 | |||
NM_001242810.2 | NP_001229739.2 | 10 | 2188 | |||
NM_001726.4 | NP_001717.3 | 10 | 1627 | |||
NM_207189.3 | NP_997072.2 | 10 | 1832 | |||
XM_006710853.3 | XP_006710916.1 | 11 | 2417 | |||
XM_006710854.3 | XP_006710917.1 | 12 | 2262 | |||
XM_006710855.3 | XP_006710918.1 | 12 | 2267 | |||
XM_006710856.3 | XP_006710919.1 | 12 | 2241 | |||
XM_006710857.3 | XP_006710920.1 | 12 | 2231 | |||
XM_011542032.2 | XP_011540334.1 | 12 | 1751 | |||
XM_011542033.2 | XP_011540335.1 | 12 | 1725 | |||
XM_011542034.2 | XP_011540336.1 | 12 | 1741 | |||
XM_011542035.2 | XP_011540337.1 | 12 | 1715 | |||
XM_011542036.2 | XP_011540338.1 | 10 | 1630 | |||
GenBank mRNA | AF019085.1 | - | 10 | 1590 | ||
AK302758.1 | - | 9 | 1436 | |||
AK303008.1 | - | 10 | 1592 | |||
AK316442.1 | - | 9 | 1499 | |||
AY338951.1 | - | 11 | 1901 | |||
BX648135.1 | - | 10 | 1923 |