Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037099.2 | NP_001032176.1 | 4 | 580 | ||
NM_001285426.1 | NP_001272355.1 | 4 | 364 | |||
NM_001285427.1 | NP_001272356.1 | 4 | 660 | |||
NM_001285428.1 | NP_001272357.1 | 3 | 289 | |||
NM_146123.3 | NP_666235.1 | 3 | 500 | |||
GenBank mRNA | AK032311.1 | - | 4 | 804 | ||
AK038633.1 | - | 4 | 616 | |||
AK043850.1 | - | 3 | 443 | |||
AK079616.1 | - | 3 | 450 | |||
AK147338.1 | - | 4 | 554 | |||
AK158606.1 | - | 3 | 291 | |||
BC026479.1 | - | 3 | 286 | 4501980 |