Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007386.2 | NP_031412.2 | 4 | 640 | ||
GenBank mRNA | AK027914.1 | - | 4 | 426 | ||
AK085951.1 | - | 4 | 389 | |||
AK129025.1 | - | 4 | 422 | |||
AK163985.1 | - | 4 | 439 | |||
BC005454.1 | - | 4 | 400 | 3494686 | ||
X61147.1 | - | 4 | 407 |