Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007386.2 | NP_031412.2 | 11, 12 | 1663 | ||
GenBank mRNA | AK027914.1 | - | 11, 12 | 1450 | ||
AK085951.1 | - | 11, 12 | 1412 | |||
AK129025.1 | - | 11, 12 | 1445 | |||
AK163985.1 | - | 11, 12 | 1462 | |||
AK207292.1 | - | 1, 2 | 116 | |||
BC005454.1 | - | 11, 12 | 1423 | 3494686 | ||
X61147.1 | - | 11, 12 | 1430 |