Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007390.3 | NP_031416.3 | 9, 10 | 1050 | ||
GenBank mRNA | AF225980.1 | - | 9, 10 | 1040 | ||
AK157809.1 | - | 9, 10 | 1047 | |||
AY574266.1 | - | 9, 10 | 1000 | |||
L37663.1 | - | 9, 10 | 1050 |