Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acrv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007391.2 | NP_031417.2 | 2 | 194 | ||
GenBank mRNA | AK005705.1 | - | 2 | 194 | ||
AK030129.1 | - | 2 | 160 | |||
BC100459.1 | - | 2 | 194 | |||
BC139101.1 | - | 2 | 153 | 8862123 | ||
BC139103.1 | - | 2 | 153 | 8862125 | ||
U31992.1 | - | 2 | 165 |