Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270996.1 | NP_001257925.1 | 22 | 3093 | ||
NM_007404.2 | NP_031430.2 | 23 | 3039 | |||
GenBank mRNA | AK122188.1 | - | 23 | 3021 | ||
AK148174.1 | - | 23 | 2925 | |||
AK155582.1 | - | 22 | 3056 | |||
AK155672.1 | - | 23 | 2933 | |||
AK161778.1 | - | 22 | 3016 | |||
BC047156.1 | - | 23 | 2917 | 6488707 | ||
BC052710.1 | - | 22 | 2965 | |||
U41765.1 | - | 23 | 2912 |