Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288578.1 | NP_001275507.1 | 6, 7 | 898 | ||
NM_007410.3 | NP_031436.2 | 6, 7 | 916 | |||
GenBank mRNA | AK002790.1 | - | 6, 7 | 838 | ||
AK076507.1 | - | 6, 7 | 845 | |||
AK146949.1 | - | 6, 7 | 860 | |||
AK159803.1 | - | 6, 7 | 896 | |||
AK180856.1 | - | 15 | ||||
BC062879.1 | - | 6, 7 | 842 | 5691327 | ||
BC090978.1 | - | 6, 7 | 839 | 30313517 | ||
M84147.1 | - | 6, 7 | 866 |