Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007422.3 | NP_031448.2 | 8 | 880 | ||
GenBank mRNA | AK004877.1 | - | 8 | 833 | ||
AK010263.1 | - | 8 | 855 | |||
AK028060.1 | - | 8 | 880 | |||
AK148420.1 | - | 8 | 862 | |||
AK170279.1 | - | 8 | 866 | |||
BC139404.1 | - | 8 | 792 | 8862426 | ||
BC139417.1 | - | 8 | 792 | 8862439 | ||
L24554.1 | - | 8 | 847 |