Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011544.1 | NP_001011544.1 | 5 | 1566 | ||
NM_005366.4 | NP_005357.2 | 5 | 1565 | |||
XM_011531164.2 | XP_011529466.1 | 4 | 2196 | |||
XM_017029521.1 | XP_016885010.1 | 7 | 1776 | |||
XM_017029522.1 | XP_016885011.1 | 7 | 3160 | |||
GenBank mRNA | AK127849.1 | - | 7 | 3053 | ||
AY747607.1 | - | 5 | 1523 | |||
BC004479.2 | - | 5 | 1501 | 3833322 |