Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177307.1 | NP_001170778.1 | 4 | 520 | ||
NM_001177308.1 | NP_001170779.1 | 3 | 393 | |||
NM_007438.4 | NP_031464.1 | 3 | 443 | |||
GenBank mRNA | AK002561.1 | - | 3 | 332 | ||
AK145639.1 | - | 3 | 330 | |||
AK154295.1 | - | 3 | 326 | |||
AK162290.1 | - | 3 | 331 | |||
BC043026.1 | - | 3 | 319 | 5691975 | ||
BC050896.1 | - | 3 | 361 | 5715436 | ||
BC066218.1 | - | 3 | 339 | 30054842 | ||
BC066801.1 | - | 3 | 339 | 30072521 | ||
BC089495.1 | - | 3 | 372 | 30312668 | ||
CF617557.1 | - | 3 | 279 | |||
CN694938.1 | - | 3 | 444 | |||
EF662059.1 | - | 3 | 374 | |||
EF662060.1 | - | 3 | 324 | |||
Y00516.1 | - | 3 | 289 |
Gene Symbol: Aldoart1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199270.1 | NP_001186199.1 | 1 | 442 | ||
GenBank mRNA | AK014956.1 | - | 1 | 443 | ||
AK016845.1 | - | 1 | 415 | |||
EF662061.1 | - | 1 | 378 |
Gene Symbol: Aldoart2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277340.1 | NP_001264269.1 | 1 | 299 | ||
GenBank mRNA | AK006425.1 | - | 1 | 286 | ||
AK016920.1 | - | 1 | 266 | |||
EF662062.1 | - | 1 | 213 |