Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008569.2 | NP_032595.2 | 5 | 770 | ||
GenBank mRNA | AK052404.1 | - | 5 | 751 | ||
AK054305.1 | - | 5 | 752 | |||
AK077847.1 | - | 5 | 771 | |||
AK090134.1 | - | 5 | 753 | |||
AK149762.1 | - | 5 | 742 | |||
AK164804.1 | - | 5 | 756 | |||
BC139002.1 | - | 5 | 751 | 8862024 | ||
BC145492.1 | - | 5 | 751 | 9054055 | ||
X80169.1 | - | 5 | 755 |