Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291890.1 | NP_001278819.1 | 12, 13 | 1264 | ||
NM_001291891.1 | NP_001278820.1 | 12, 13 | 1459 | |||
NM_009616.4 | NP_033746.1 | 12, 13 | 1449 | |||
GenBank mRNA | AF019887.2 | - | 12, 13 | 1354 | ||
AK147217.1 | - | 12, 13 | 1372 | |||
AK147549.1 | - | 12, 13 | 1265 | |||
BC093480.1 | - | 12, 13 | 1450 | |||
D50410.2 | - | 12, 13 | 1381 |