Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009655.2 | NP_033785.1 | 9 | 1572 | ||
GenBank mRNA | AB003026.1 | - | 9 | 1234 | ||
AK030851.1 | - | 9 | 1573 | |||
AK031391.1 | - | 2648 | ||||
BC027280.1 | - | 9 | 1229 | 3501215 | ||
L25274.1 | - | 4 | 364 | |||
U95030.1 | - | 9 | 1040 |