Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G3bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF145285.1 | - | 12 | 1821 | ||
AK076117.1 | - | 11 | 1772 | |||
AK145697.1 | - | 12 | 1870 | |||
AK149427.1 | - | 12 | 1849 | |||
AK151732.1 | - | 11 | 1807 | |||
AK151965.1 | - | 11 | 1736 | |||
AK152792.1 | - | 11 | 1740 | |||
AK153221.1 | - | 12 | 1861 | |||
AK153893.1 | - | 12 | 1809 | |||
AK167186.1 | - | 11 | 1777 | |||
AK169571.1 | - | 1 | 331 | |||
AK172256.1 | - | 11 | 1742 | |||
AK220349.1 | - | 1 | 469 | |||
BC048176.1 | - | 11 | 1759 | 6825491 | ||
U65313.1 | - | 11 | 1723 |