Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il27ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016671.3 | NP_057880.2 | 10, 11 | 1432 | ||
GenBank mRNA | AF053005.1 | - | 10, 11 | 1400 | ||
AF265243.1 | - | 10, 11 | 1411 | |||
AK040457.1 | - | 8, 9 | 1772 | |||
AK133753.1 | - | 10, 11 | 1447 | |||
BC032878.1 | - | 10, 11 | 1432 | 1244998 |