Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acadvl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017366.3 | NP_059062.1 | 7 | 663 | ||
GenBank mRNA | AF017176.1 | - | 4 | 311 | ||
AK076037.1 | - | 7 | 624 | |||
AK146334.1 | - | 2 | 136 | |||
AK196376.1 | - | 2 | 226 | |||
AK215457.1 | - | 2 | 226 | |||
BC026559.1 | - | 7 | 642 | 5037878 | ||
Z71189.1 | - | 7 | 584 |