Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAALADL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005468.2 | NP_005459.2 | 4 | 564 | ||
XM_011544706.2 | XP_011543008.1 | 4 | 884 | |||
XM_011544707.2 | XP_011543009.1 | 5 | 532 | |||
XM_011544708.2 | XP_011543010.1 | 4 | 884 | |||
XM_011544709.2 | XP_011543011.1 | 4 | 904 | |||
XM_011544710.2 | XP_011543012.1 | 4 | 901 | |||
XM_011544711.2 | XP_011543013.1 | 4 | 906 | |||
XM_011544712.2 | XP_011543014.1 | 4 | 907 | |||
GenBank mRNA | AJ012371.1 | - | 4 | 564 |