Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACOT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005469.3 | NP_005460.2 | 1, 2 | 219 | ||
XM_011528480.2 | XP_011526782.1 | 1, 2 | 181 | |||
GenBank mRNA | AF014404.1 | - | 1, 2 | 155 | ||
AF124264.1 | - | 1, 2 | 125 | |||
AK296973.1 | - | 1, 2 | 207 | |||
BC117155.1 | - | 1, 2 | 147 | 40125706 | ||
BC117157.1 | - | 1, 2 | 147 | 40125708 | ||
BI911038.1 | - | 1, 2 | 205 | |||
CR541771.1 | - | 1, 2 | 125 | |||
CR541778.1 | - | 1, 2 | 125 | |||
DB470803.3 | - | 1, 2 | 219 | |||
X86032.1 | - | 1, 2 | 193 |