Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rangrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285441.1 | NP_001272370.1 | 5 | 537 | ||
NM_001285442.1 | NP_001272371.1 | 5 | 612 | |||
NM_001285443.1 | NP_001272372.1 | 5 | 624 | |||
NM_021329.3 | NP_067304.1 | 5 | 633 | |||
GenBank mRNA | AF243512.1 | - | 5 | 604 | ||
AK010288.1 | - | 5 | 633 | |||
BB618250.1 | - | 5 | 631 | |||
BC018290.1 | - | 5 | 536 | |||
BC038470.1 | - | 5 | 614 | 5344659 | ||
BC087921.1 | - | 5 | 598 | 6772909 | ||
BX523534.1 | - | 2 | 138 | |||
CF751505.1 | - | 5 | 465 |