Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163028.1 | NP_001156500.1 | 3, 4 | 499 | ||
NM_021486.3 | NP_067461.2 | 3, 4 | 499 | |||
GenBank mRNA | AF271298.1 | - | 3, 4 | 530 | ||
AF294899.1 | - | 3, 4 | 512 | |||
AJ278064.2 | - | 3, 4 | 511 | |||
AK054171.1 | - | 2, 3 | 251 | |||
AK146472.1 | - | 3, 4 | 496 | |||
BC125328.1 | - | 3, 4 | 366 | 40129843 | ||
BC125330.1 | - | 3, 4 | 366 | 40129845 | ||
BC145036.1 | - | 3, 4 | 366 |