Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024213.2 | NP_077175.1 | 3, 4 | 502 | ||
GenBank mRNA | AK088759.1 | - | 3, 4 | 492 | ||
AK146000.1 | - | 3, 4 | 499 | |||
AK148625.1 | - | 3, 4 | 511 | |||
AK166061.1 | - | 3, 4 | 625 | |||
AK168031.1 | - | 3, 4 | 499 | |||
AK168897.1 | - | 4, 5 | 584 | |||
BC016237.1 | - | 3, 4 | 485 | 4504779 | ||
BC024870.1 | - | 3, 4 | 502 | 4949079 |