Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025394.3 | NP_079670.1 | 2, 3 | 219 | ||
GenBank mRNA | AK003864.1 | - | 2, 3 | 182 | ||
AK008581.1 | - | 2, 3 | 136 | |||
BC055352.1 | - | 2, 3 | 197 | 5011269 | ||
BE647192.1 | - | 1, 2 | 21 | |||
BU555283.1 | - | 2, 3 | 222 | |||
BY773505.1 | - | 2, 3 | 212 |