Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025836.3 | NP_080112.1 | 8 | 1632 | ||
GenBank mRNA | AK003971.1 | - | 1 | 484 | ||
AK004970.1 | - | 8 | 1634 | |||
AK012517.1 | - | 8 | 1632 | |||
AK132313.1 | - | 1633 | ||||
AK167214.1 | - | 8 | 1632 | |||
AK210643.1 | - | 1 | 319 |