Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290321.2 | NP_001277250.1 | 2 | 645 | ||
NM_001290322.2 | NP_001277251.1 | 2 | 645 | |||
NM_005509.5 | NP_005500.4 | 2 | 645 | |||
XM_005271909.4 | XP_005271966.1 | 3 | 343 | |||
XM_005271910.4 | XP_005271967.1 | 2 | 688 | |||
XM_005271912.2 | XP_005271969.1 | 3 | 327 | |||
XM_011543212.2 | XP_011541514.1 | 2 | 688 | |||
XM_011543213.2 | XP_011541515.1 | 3 | 273 | |||
XM_017009141.1 | XP_016864630.1 | 3 | 343 | |||
XM_017009142.1 | XP_016864631.1 | 3 | 273 | |||
XM_017009143.1 | XP_016864632.1 | 4 | 522 | |||
XM_017009144.1 | XP_016864633.1 | 2 | 225 | |||
XM_017009145.1 | XP_016864634.1 | 2 | 95 | |||
XM_017009146.1 | XP_016864635.1 | 2 | 225 | |||
XM_017009147.1 | XP_016864636.1 | 2 | 688 | |||
XM_017009148.1 | XP_016864637.1 | 2 | 624 | |||
GenBank mRNA | AB593142.1 | - | 2 | 645 | ||
AJ005821.1 | - | 2 | 242 | |||
BC028004.1 | - | 3 | 230 | |||
BC144537.1 | - | 2 | 188 | 9053064 | ||
BC150616.1 | - | 2 | 188 | 9056986 |