Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nvl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026171.1 | NP_080447.1 | 4, 5 | 334 | ||
GenBank mRNA | AK004676.1 | - | 4, 5 | 366 | ||
AK013982.1 | - | 4, 5 | 333 | |||
AK018049.1 | - | 4, 5 | 330 | |||
AK140509.1 | - | 4, 5 | 365 | |||
BC031847.1 | - | 4, 5 | 317 | 3964470 |