Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026390.2 | NP_080666.2 | 2 | 391 | ||
GenBank mRNA | AK011233.1 | - | 2 | 288 | ||
AK012824.1 | - | 2 | 285 | |||
AK019371.1 | - | 2 | 284 | |||
AK029431.1 | - | 2 | 437 | |||
AK032206.1 | - | 2 | 270 | |||
AK075716.1 | - | 2 | 126 | |||
AK147795.1 | - | 2 | 264 | |||
AK148333.1 | - | 2 | 277 | |||
BC012271.1 | - | 2 | 403 | |||
BC019795.1 | - | 2 | 410 | 4158848 | ||
BC030190.1 | - | 2 | 400 |