Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026850.4 | NP_081126.2 | 6 | 975 | ||
GenBank mRNA | AK004334.1 | - | 6 | 970 | ||
AK078372.1 | - | 6 | 976 | |||
AK168509.1 | - | 6 | 1016 | |||
AK169572.1 | - | 6 | 967 | |||
BC005601.1 | - | 6 | 997 | 3483084 |