Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lzic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026963.5 | NP_081239.1 | 7 | 1227 | ||
GenBank mRNA | AK007657.1 | - | 7 | 716 | ||
AK146824.1 | - | 7 | 1294 | |||
BC023174.1 | - | 7 | 1227 | 4952948 | ||
DQ058008.1 | - | 6 | 552 |