Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027494.3 | NP_081770.3 | 3 | 408 | ||
GenBank mRNA | AK017857.1 | - | 3 | 304 | ||
AK032349.1 | - | 3 | 386 | |||
AK033469.1 | - | 3 | 358 | |||
AK156770.1 | - | 3 | 380 | |||
AK166990.1 | - | 3 | 315 | |||
AK167126.1 | - | 3 | 313 | |||
AK167736.1 | - | 3 | 315 | |||
BC013555.1 | - | 2 | 60 | 4238634 |