Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024918.1 | NP_001020089.1 | 6 | 920 | ||
NM_027689.3 | NP_081965.2 | 2 | 252 | |||
GenBank mRNA | AB086957.1 | - | 6 | 774 | ||
AB089184.1 | - | 2 | 253 | |||
AK016791.1 | - | 2 | 253 | |||
AK034131.1 | - | 6 | 907 | |||
AK132673.1 | - | 3 | 315 | |||
AY102010.2 | - | 6 | 803 | |||
AY342003.1 | - | 6 | 785 | |||
BC138719.1 | - | 2 | 240 | 8861741 | ||
BC138720.1 | - | 2 | 240 | 8861742 |