Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Susd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027890.5 | NP_082166.3 | 3 | 432 | ||
GenBank mRNA | AK004703.2 | - | 3 | 402 | ||
AK028817.1 | - | 3 | 408 | |||
AK037059.1 | - | 3 | 414 | |||
AK037127.1 | - | 3 | 383 | |||
AK038944.1 | - | 3 | 433 | |||
AK042469.1 | - | 3 | 416 | |||
AK128959.1 | - | 3 | 404 | |||
AK167155.1 | - | 3 | 433 | |||
BB632717.1 | - | 3 | 433 | |||
BC026837.1 | - | 3 | 385 | 4222578 | ||
BC066833.1 | - | 1 | 50 |