Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028031.3 | NP_082307.1 | 3 | 306 | ||
GenBank mRNA | AK010382.1 | - | 3 | 306 | ||
AK049640.1 | - | 2 | 136 | |||
AK145855.1 | - | 3 | 226 | |||
AK146196.1 | - | 3 | 278 | |||
BC019493.1 | - | 3 | 225 | |||
BC046599.1 | - | 3 | 225 | 6311721 |